N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Hun-Soo Byun.; Nam-Seok Jeon. Phase behavior measurement of the binary carbon dioxide-N,N-dimethylacetamide and carbon dioxide-N,N-diethylacetamide systems at high pressure. Fluid Phase Equilib. 2000, 167 (1),113-122 .
Allan F. Sowinski.; George M. Whitesides. SN2 displacements and reductive coupling of ketones with olefins in N,N-diethylacetamide and N-ethylpyrrolidone.J. Org. Chem. 1979, 44 (14),2369-2376 .