Thermo Scientific Chemicals

N,N-Diethylacetamide, 99%, Thermo Scientific Chemicals

Catalog number: B20762.36
500 g, Each
Thermo Scientific Chemicals

N,N-Diethylacetamide, 99%, Thermo Scientific Chemicals

Catalog number: B20762.36
500 g, Each
Quantity
Catalog number: B20762.36
also known as B20762-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
685-91-6
IUPAC Name
N,N-diethylacetamide
Molecular Formula
C6H13NO
InChI Key
AJFDBNQQDYLMJN-UHFFFAOYSA-N
SMILES
CCN(CC)C(C)=O
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4375-1.4415 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms

Description

N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text