Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals
Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20765.14
also known as B20765-14
Price (USD)/ Each
320.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
320.00
Add to cart
Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals
Catalog numberB20765.14
Price (USD)/ Each
320.00
-
Add to cart
Chemical Identifiers
CAS18856-68-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear yellow
Assay (GC)≥92.5%
Refractive Index1.5040-1.5100 @ 20°C
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Clémentine Féau.;Emmanuel Klein.; Paul Kerth.; Luc Lebeau. Preparation and Optical Properties of Novel 3-Alkoxycarbonyl Aza- and Diazacoumarins.Synth. Commun. 2010, 40 (20),3033-3045 .