Thermo Scientific Chemicals

2,4,4-Trimethyl-2-pentene, 97%, Thermo Scientific Chemicals

Catalog number: B20773.14
25 g, Each
Thermo Scientific Chemicals

2,4,4-Trimethyl-2-pentene, 97%, Thermo Scientific Chemicals

Catalog number: B20773.14
25 g, Each
Quantity
Catalog number: B20773.14
also known as B20773-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-40-4
IUPAC Name
2,4,4-trimethylpent-2-ene
Molecular Formula
C8H16
InChI Key
LAAVYEUJEMRIGF-UHFFFAOYSA-N
SMILES
CC(C)=CC(C)(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4135-1.4185 @ 20?C
Assay (GC)
≥96.0%

Description

2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text