Thermo Scientific Chemicals

Diethylene glycol mono-n-hexyl ether, 97%, Thermo Scientific Chemicals

Catalog number: B20774.0F
2500 mL, Each
Thermo Scientific Chemicals

Diethylene glycol mono-n-hexyl ether, 97%, Thermo Scientific Chemicals

Catalog number: B20774.0F
2500 mL, Each
Quantity
Catalog number: B20774.0F
also known as B20774-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-59-4
IUPAC Name
2-[2-(hexyloxy)ethoxy]ethan-1-ol
Molecular Formula
C10H22O3
InChI Key
GZMAAYIALGURDQ-UHFFFAOYSA-N
SMILES
CCCCCCOCCOCCO
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Refractive Index
1.4345-1.4395 @ 20?C
Form
Liquid

Description

Used as a solvent for coatings and degreasing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a solvent for coatings and degreasing.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text