Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number B20780.06
also known as B20780-06
Price (USD)/ Each
144.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
144.00
Add to cart
Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB20780.06
Price (USD)/ Each
144.00
-
Add to cart
Chemical Identifiers
CAS96232-71-2
IUPAC Name(2Z)-4,5-dibromo-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Molecular FormulaC6H4Br2O3S
InChI KeyYZPNGCXFJCPJSV-XQRVVYSFSA-N
SMILESCOC(O)=C1/SC(Br)=C(Br)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)124.5-133.5?C
Appearance (Color)Pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only