Thermo Scientific Chemicals

1-Iodopentane, 98%, stab with copper, Thermo Scientific Chemicals

Catalog number: B20781.36
500 g, Each
Thermo Scientific Chemicals

1-Iodopentane, 98%, stab with copper, Thermo Scientific Chemicals

Catalog number: B20781.36
500 g, Each
Quantity
Catalog number: B20781.36
also known as B20781-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-17-1
IUPAC Name
1-iodopentane
Molecular Formula
C5H11I
InChI Key
BLXSFCHWMBESKV-UHFFFAOYSA-N
SMILES
CCCCCI
Appearance (Color)
Clear colorless to yellow or pale orange
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4935-1.4975 @ 20°C

Description

1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text