Thermo Scientific Chemicals

2-Cyclohexylethanol, 99%, Thermo Scientific Chemicals

Catalog number: B20782.30
250 g, Each
Thermo Scientific Chemicals

2-Cyclohexylethanol, 99%, Thermo Scientific Chemicals

Catalog number: B20782.30
250 g, Each
Quantity
Catalog number: B20782.30
also known as B20782-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4442-79-9
IUPAC Name
2-cyclohexylethan-1-ol
Molecular Formula
C8H16O
InChI Key
QJQZRLXDLORINA-UHFFFAOYSA-N
SMILES
OCCC1CCCCC1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4645-1.4675 @ 20?C

Description

Used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent.

Solubility
Soluble in alcohol and dipropylene glycol. Soluble in water (1750 mg/L at 25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text