1,4-Cyclohexanedione bis(ethylene acetal), 99%, Thermo Scientific Chemicals
1,4-Cyclohexanedione bis(ethylene acetal), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Cyclohexanedione bis(ethylene acetal), 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
Catalog number B20784.09
also known as B20784-09
Price (USD)/ Each
46.80
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
46.80
Add to cart
1,4-Cyclohexanedione bis(ethylene acetal), 99%, Thermo Scientific Chemicals
Catalog numberB20784.09
Price (USD)/ Each
46.80
-
Add to cart
Chemical Identifiers
CAS183-97-1
IUPAC Name1,4,9,12-tetraoxadispiro[4.2.4⁸.2⁵]tetradecane
Molecular FormulaC10H16O4
InChI KeyYSMVSEYPOBXSOK-UHFFFAOYSA-N
SMILESC1COC2(CCC3(CC2)OCCO3)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)76.5-82.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only