Thermo Scientific Chemicals

Aluminum trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: B20785.14
25 g, Each
Thermo Scientific Chemicals

Aluminum trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: B20785.14
25 g, Each
Quantity
Catalog number: B20785.14
also known as B20785-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74974-61-1
IUPAC Name
aluminium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3AlF9O9S3
InChI Key
FKOASGGZYSYPBI-UHFFFAOYSA-K
SMILES
[Al+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Appearance (Color)
White
Assay (unspecified)
≥98.5 to ≤101.5% based on Fluorine EA
Elemental Analysis
Fluorine F: 35.52-36.60% (Theory: 36.06%)
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text