Thermo Scientific Chemicals

N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals

Catalog number: B20805.22
100 g, Each
Thermo Scientific Chemicals

N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals

Catalog number: B20805.22
100 g, Each
Quantity
Catalog number: B20805.22
also known as B20805-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2387-23-7
IUPAC Name
1,3-dicyclohexylurea
Molecular Formula
C13H24N2O
InChI Key
ADFXKUOMJKEIND-UHFFFAOYSA-N
SMILES
O=C(NC1CCCCC1)NC1CCCCC1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
226.0-235.0?C
Assay (HPLC)
≥97.5%
Appearance (Color)
White to pale cream to pale brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text