5-Fluoroisatin, 98%, Thermo Scientific Chemicals
5-Fluoroisatin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoroisatin, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B20831.14
also known as B20831-14
Price (USD)/ Each
239.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
239.00
Add to cart
5-Fluoroisatin, 98%, Thermo Scientific Chemicals
Catalog numberB20831.14
Price (USD)/ Each
239.00
-
Add to cart
Chemical Identifiers
CAS443-69-6
IUPAC Name5-fluoro-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4FNO2
InChI KeyGKODDAXOSGGARJ-UHFFFAOYSA-N
SMILESFC1=CC=C2NC(=O)C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to dark brown
Formpowder
Assay (HPLC)> 97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only