Furazolidone, 98%, Thermo Scientific Chemicals
Furazolidone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Furazolidone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B20834.14
also known as B20834-14
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
Furazolidone, 98%, Thermo Scientific Chemicals
Catalog numberB20834.14
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS67-45-8
IUPAC Name3-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]-1,3-oxazolidin-2-one
Molecular FormulaC8H7N3O5
InChI KeyPLHJDBGFXBMTGZ-WEVVVXLNSA-N
SMILES[O-][N+](=O)C1=CC=C(O1)C=NN1CCOC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Assay (HPLC)≥97.5%
Monoamine oxidase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only