Thermo Scientific Chemicals

Furazolidone, 98%, Thermo Scientific Chemicals

Catalog number: B20834.22
100 g, Each
Thermo Scientific Chemicals

Furazolidone, 98%, Thermo Scientific Chemicals

Catalog number: B20834.22
100 g, Each
Quantity
Catalog number: B20834.22
also known as B20834-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-45-8
IUPAC Name
3-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]-1,3-oxazolidin-2-one
Molecular Formula
C8H7N3O5
InChI Key
PLHJDBGFXBMTGZ-WEVVVXLNSA-N
SMILES
[O-][N+](=O)C1=CC=C(O1)C=NN1CCOC1=O
Form
Powder
Appearance (Color)
Yellow
Assay (HPLC)
≥97.5%

Description

Monoamine oxidase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text