2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific Chemicals
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20840.06
also known as B20840-06
Price (USD)/ Each
63.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
63.00
Add to cart
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific Chemicals
Catalog numberB20840.06
Price (USD)/ Each
63.00
-
Add to cart
Chemical Identifiers
CAS94651-33-9
IUPAC Name2-(trifluoromethoxy)benzaldehyde
Molecular FormulaC8H5F3O2
InChI KeyCPHXLFKIUVVIOQ-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Free acid (titration)≤2%
Assay (GC)≥95.0%
Refractive Index1.4515-1.4575 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only