Thermo Scientific Chemicals

Cyclooctanone, 98%, Thermo Scientific Chemicals

Catalog number: B20911.22
100 g, Each
Thermo Scientific Chemicals

Cyclooctanone, 98%, Thermo Scientific Chemicals

Catalog number: B20911.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B20911.22
also known as B20911-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
502-49-8
IUPAC Name
cyclooctanone
Molecular Formula
C8H14O
InChI Key
IIRFCWANHMSDCG-UHFFFAOYSA-N
SMILES
O=C1CCCCCCC1
Appearance (Color)
White to pale cream
Assay (GC)
≥97.5%
Form
Crystalline or fused solid
Identification (FTIR)
Conforms

Description

Cyclooctanone is a aliphatic cycle that exhibited inhibitory activity towards aldosterone synthase, a promising therapeutic target for the treatment of cardiovascular diseases related to abnormally high aldosterone level. It is also used in the synthesis of 14-membered lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 20°C 15g/L. Soluble in acetone, alcohol, chloroform, methanol and benzene.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text