Thermo Scientific Chemicals

3-Methoxyphenylacetone, 97%, Thermo Scientific Chemicals

Catalog number: B20916.06
5 g, Each
Thermo Scientific Chemicals

3-Methoxyphenylacetone, 97%, Thermo Scientific Chemicals

Catalog number: B20916.06
5 g, Each
Quantity
Catalog number: B20916.06
also known as B20916-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3027-13-2
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear pale yellow to yellow
Refractive Index
1.5220-1.5270 @ 20?C

Description

3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text