2-(4-Methylphenyl)ethanol, 99%, Thermo Scientific Chemicals
2-(4-Methylphenyl)ethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(4-Methylphenyl)ethanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20918.06
also known as B20918-06
Price (USD)/ Each
105.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
105.00
Add to cart
2-(4-Methylphenyl)ethanol, 99%, Thermo Scientific Chemicals
Catalog numberB20918.06
Price (USD)/ Each
105.00
-
Add to cart
Chemical Identifiers
CAS699-02-5
IUPAC Name2-(4-methylphenyl)ethan-1-ol
Molecular FormulaC9H12O
InChI KeyDAVFJRVIVZOKKS-UHFFFAOYSA-N
SMILESCC1=CC=C(CCO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5255-1.5285 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only