5-(2,3-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals
5-(2,3-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(2,3-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number B20921.06
also known as B20921-06
Price (USD)/ Each
624.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
624.00
Add to cart
5-(2,3-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals
Catalog numberB20921.06
Price (USD)/ Each
624.00
-
Add to cart
Chemical Identifiers
CAS175205-12-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream to cream to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water 0.026 g/L (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only