1,2-Dibromo-4-fluorobenzene, 98%, Thermo Scientific Chemicals
1,2-Dibromo-4-fluorobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dibromo-4-fluorobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Catalog number B20935.09
also known as B20935-09
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
1,2-Dibromo-4-fluorobenzene, 98%, Thermo Scientific Chemicals
Catalog numberB20935.09
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS2369-37-1
IUPAC Name1,2-dibromo-4-fluorobenzene
Molecular FormulaC6H3Br2F
InChI KeyRNTGKISRXVFIIP-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5770-1.5810 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only