2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%, Thermo Scientific Chemicals
2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20936.14
also known as B20936-14
Price (USD)/ Each
225.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
225.00
Add to cart
2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%, Thermo Scientific Chemicals
Catalog numberB20936.14
Price (USD)/ Each
225.00
-
Add to cart
Chemical Identifiers
CAS4383-08-8
IUPAC Name2-methyl-1-phenylprop-2-en-1-ol
Molecular FormulaC10H12O
InChI KeyZGYBYYJGIKPBFD-UHFFFAOYNA-N
SMILESCC(=C)C(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>85.0%
Refractive Index1.5690-1.5720
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only