2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B20938.06
also known as B20938-06
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Catalog numberB20938.06
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS57729-79-0
IUPAC Name2-chloro-6-nitro-4-(trifluoromethyl)aniline
Molecular FormulaC7H4ClF3N2O2
InChI KeyJLWRJMVXRUKFPA-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=C(C=C1[N+]([O-])=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)61.0-70.0?C
Assay (GC)≥96.0%
Appearance (Color)Orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only