Thermo Scientific Chemicals

3,5-Dichlorosalicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B20941.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dichlorosalicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B20941.06
5 g, Each
Quantity
Catalog number: B20941.06
also known as B20941-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-60-8
IUPAC Name
3,5-dichloro-2-hydroxybenzaldehyde
Molecular Formula
C7H4Cl2O2
InChI Key
FABVMBDCVAJXMB-UHFFFAOYSA-N
SMILES
OC1=C(Cl)C=C(Cl)C=C1C=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream to yellow
Assay (GC)
≥97.5%
Melting Point (clear melt)
91.0-97.0?C

Description

3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.

Solubility
Insoluble in water. Solubility in methanol is almost transparent.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text