3,5-Dichlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
3,5-Dichlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dichlorosalicylaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B20941.14
also known as B20941-14
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
3,5-Dichlorosalicylaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberB20941.14
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS90-60-8
IUPAC Name3,5-dichloro-2-hydroxybenzaldehyde
Molecular FormulaC7H4Cl2O2
InChI KeyFABVMBDCVAJXMB-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=C(Cl)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Pale cream to cream to yellow
Assay (GC)≥97.5%
Melting Point (clear melt)91.0-97.0?C
3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.

Solubility
Insoluble in water. Solubility in methanol is almost transparent.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Takashiro Akitsu, et al. Racemic 3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.Acta Crystallogr C.,2004,60(11), o801-o802.
  2. Yongming Cui, et al. Synthesis, structures and urease inhibition studies of Schiff base metal complexes derived from 3,5-dibromosalicylaldehyde.Eur J Med Chem.,2012,58323-331.