5-Chloro-2-fluoroaniline, 97%, Thermo Scientific Chemicals
5-Chloro-2-fluoroaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-2-fluoroaniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number B20951.18
also known as B20951-18
Price (USD)/ Each
262.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
262.00
Add to cart
5-Chloro-2-fluoroaniline, 97%, Thermo Scientific Chemicals
Catalog numberB20951.18
Price (USD)/ Each
262.00
-
Add to cart
Chemical Identifiers
CAS2106-05-0
IUPAC Name5-chloro-2-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyJCYROOANFKVAIB-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5575-1.5625 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only