4-Carboxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-Carboxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Carboxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B20954.06
also known as B20954-06
Price (USD)/ Each
129.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
129.00
Add to cart
4-Carboxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberB20954.06
Price (USD)/ Each
129.00
-
Add to cart
Chemical Identifiers
CAS14047-29-1
IUPAC Name4-(dihydroxyboranyl)benzoic acid
Molecular FormulaC7H7BO4
InChI KeySIAVMDKGVRXFAX-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0%
FormPowder or lumps or granules
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only