1,4-Diacetoxybenzene, 98%, Thermo Scientific Chemicals
1,4-Diacetoxybenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Diacetoxybenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
250 g
Catalog number B20987.22
also known as B20987-22
Price (USD)/ Each
91.20
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
91.20
Add to cart
1,4-Diacetoxybenzene, 98%, Thermo Scientific Chemicals
Catalog numberB20987.22
Price (USD)/ Each
91.20
-
Add to cart
Chemical Identifiers
CAS1205-91-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)117.0-124.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only