Thermo Scientific Chemicals

Bis(diethylamino)chlorophosphine, 94%, Thermo Scientific Chemicals

Catalog number: B21019.14
25 g, Each
Thermo Scientific Chemicals

Bis(diethylamino)chlorophosphine, 94%, Thermo Scientific Chemicals

Catalog number: B21019.14
25 g, Each
Quantity
Catalog number: B21019.14
also known as B21019-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
685-83-6
IUPAC Name
[chloro(diethylamino)phosphanyl]diethylamine
Molecular Formula
C8H20ClN2P
InChI Key
JVEHJSIFWIIFHM-UHFFFAOYSA-N
SMILES
CCN(CC)P(Cl)N(CC)CC
Phosphorus-31 NMR
Conforms to structure
Form
Liquid
Proton NMR
Conforms to structure
Appearance (Color)
Clear colorless
Assay by NMR
≥92.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text