Thermo Scientific Chemicals

Tetrakis(trimethylsilyl)silane, 98%, Thermo Scientific Chemicals

Catalog number: B21020.03
1 g, Each
Thermo Scientific Chemicals

Tetrakis(trimethylsilyl)silane, 98%, Thermo Scientific Chemicals

Catalog number: B21020.03
1 g, Each
Quantity
Catalog number: B21020.03
also known as B21020-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4098-98-0
IUPAC Name
1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane
Molecular Formula
C12H36Si5
InChI Key
BOJSDHZZKKYWAS-UHFFFAOYSA-N
SMILES
C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C
Assay (GC)
≥97.5%
Form
Powder
Appearance (Color)
White to pale cream
Water Content (Karl Fischer Titration)
≤1.0%

Description

It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text