Thermo Scientific Chemicals

2-Ethyl-1-butanol, 99%, Thermo Scientific Chemicals

Catalog number: B21026.0F
2500 mL, Each
Thermo Scientific Chemicals

2-Ethyl-1-butanol, 99%, Thermo Scientific Chemicals

Catalog number: B21026.0F
2500 mL, Each
Quantity
Catalog number: B21026.0F
also known as B21026-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-95-0
IUPAC Name
2-ethylbutan-1-ol
Molecular Formula
C6H14O
InChI Key
TZYRSLHNPKPEFV-UHFFFAOYSA-N
SMILES
CCC(CC)CO
Form
Liquid
Refractive Index
1.4205-1.4235 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.

Solubility
Soluble in water (4 mg/ml at 25°C), alcohol, ether, and most organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text