Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: B21034.MD
250 mg, Each
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: B21034.MD
250 mg, Each
Quantity
Catalog number: B21034.MD
also known as B21034-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7240-90-6
IUPAC Name
X-gal
Molecular Formula
C23H41Cu2N5O5
InChI Key
GGILAJWFDJWIQO-UHFFFAOYSA-N
SMILES
[Cu+].[Cu++].CC(O)O.CC(O)O.CN1CCN(CC2=CC(=CC(CN3CCN(C)CC3)=C2O)C#N)CC1
Form
Powder
Optical Rotation
-61.6 ± 0.1? (C=1 in DMF:H2O 50:50)
Appearance (Color)
White
Assay (HPLC)
≥98.0%

Description

It is a substrate for beta-galactosidase, which cleaves the glycosidic bond to give 5-bromo-4-chloro-3-hydroxy-1H-indole, which immediately dimerises to give an intensely blue product. It is also used to detect activity of this enzyme in histochemistry and bacteriology. It is employed as a colorless, endogenous or exogenous pigment precursors that may be transformed by biological mechanisms into colored compounds. They are used in biochemical assays and in diagnosis as indicators, particularly in the form of enzyme substrates. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate for beta-galactosidase, which cleaves the glycosidic bond to give 5-bromo-4-chloro-3-hydroxy-1H-indole, which immediately dimerises to give an intensely blue product. It is also used to detect activity of this enzyme in histochemistry and bacteriology. It is employed as a colorless, endogenous or exogenous pigment precursors that may be transformed by biological mechanisms into colored compounds. They are used in biochemical assays and in diagnosis as indicators, particularly in the form of enzyme substrates. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Solubility
Soluble in 100 mM in DMSO.

Notes
Moisture & Light sensitive. Store away from oxidizing agents, heat, water/moisture and light. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text