Thermo Scientific Chemicals

Isobutyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: B21048.36
500 g, Each
Thermo Scientific Chemicals

Isobutyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: B21048.36
500 g, Each
Quantity
Catalog number: B21048.36
also known as B21048-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18395-30-7
IUPAC Name
trimethoxy(2-methylpropyl)silane
Molecular Formula
C7H18O3Si
InChI Key
XYJRNCYWTVGEEG-UHFFFAOYSA-N
SMILES
CO[Si](CC(C)C)(OC)OC
Assay (GC)
≥96.0%
Refractive Index
1.3935-1.3985 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text