Thermo Scientific Chemicals

N-Fmoc-glycine, 98%, Thermo Scientific Chemicals

Catalog number: B21050.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-glycine, 98%, Thermo Scientific Chemicals

Catalog number: B21050.06
5 g, Each
Quantity
Catalog number: B21050.06
also known as B21050-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29022-11-5
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
Molecular Formula
C17H15NO4
InChI Key
NDKDFTQNXLHCGO-UHFFFAOYSA-N
SMILES
OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Form
Powder
Comment
Material sourced either in [UK] or [US]
Proton NMR
Conforms [US]
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
171.0-178.0?C [UK]

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text