Thermo Scientific Chemicals

Chlorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: B21052.AP
500 mL, Each
Thermo Scientific Chemicals

Chlorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: B21052.AP
500 mL, Each
Quantity
Catalog number: B21052.AP
also known as B21052-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-90-7
IUPAC Name
chlorobenzene
Molecular Formula
C6H5Cl
InChI Key
MVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.5230-1.5260 @ 20?C
Assay (GC)
≥98.5%

Description

Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

Solubility
Miscible with alcohol, chloroform, diethyl ether and benzene. Slightly immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text