Thermo Scientific Chemicals

1,5-Bis(diphenylphosphino)pentane, 97%, Thermo Scientific Chemicals

Catalog number: B21054.03
1 g, Each
Thermo Scientific Chemicals

1,5-Bis(diphenylphosphino)pentane, 97%, Thermo Scientific Chemicals

Catalog number: B21054.03
1 g, Each
Quantity
Catalog number: B21054.03
also known as B21054-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27721-02-4
IUPAC Name
[5-(diphenylphosphanyl)pentyl]diphenylphosphane
Molecular Formula
C29H30P2
InChI Key
MZFPAWGWFDGCHP-UHFFFAOYSA-N
SMILES
C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Powder or crystals or crystalline powder or chunks
Assay from Supplier's CofA
≥96.0% (U.S. specification)
Elemental Analysis
H: 6.58-7.13% (Theory: 6.86%) (non-U.S. specification)
Elemental Analysis
C: 75.91-82.24% (Theory: 79.08%) (non-U.S. specification)

Description

1,5-Bis(diphenylphosphino)pentane is used as a ligand for the cross coupling reactions. It is used in the manufacturing of glass bottle PTFE bottle, aluminum foil bag and in cardboard drum. combination of a 1,5-bis(diphenylphosphino)pentane ligand and the addition of N,N,N?,N?-tetramethylethylenediamine (TMEDA) as a co-catalyst are the key factors in obtaining the corresponding aryl nitriles with improved catalyst productivities and selectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Bis(diphenylphosphino)pentane is used as a ligand for the cross coupling reactions. It is used in the manufacturing of glass bottle PTFE bottle, aluminum foil bag and in cardboard drum. combination of a 1,5-bis(diphenylphosphino)pentane ligand and the addition of N,N,N′,N′-tetramethylethylenediamine (TMEDA) as a co-catalyst are the key factors in obtaining the corresponding aryl nitriles with improved catalyst productivities and selectivities.

Solubility
Soluble in N,N-dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text