Thermo Scientific Chemicals

2-Methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21071.06
5 g, Each
Thermo Scientific Chemicals

2-Methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21071.06
5 g, Each
Quantity
Catalog number: B21071.06
also known as B21071-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5720-06-9
IUPAC Name
(2-methoxyphenyl)boronic acid
Molecular Formula
C7H9BO3
InChI Key
ROEQGIFOWRQYHD-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1B(O)O
Assay (Aqueous acid-base Titration)
≥96.0%
Assay (HPLC)
≥96.0%
Proton NMR
Conforms to structure
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder

Description

2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text