4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%, Thermo Scientific Chemicals
4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number B21075.01
also known as B21075-01
Price (USD)/ Each
51.30
-
Add to cart
Quantity:
0.25 g
Request bulk or custom format
Price (USD)/ Each
51.30
Add to cart
4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%, Thermo Scientific Chemicals
Catalog numberB21075.01
Price (USD)/ Each
51.30
-
Add to cart
Chemical Identifiers
CAS6160-78-7
IUPAC Name4-methyl-7-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC16H18O8
InChI KeyYUDPTGPSBJVHCN-DZQJYWQESA-N
SMILESCC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥97.0%
It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store at -20°C. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Manafi M.; Kneifel W. A combined chromogenic-fluorogenic medium for the simultaneous detection of coliform groups and E. coli in water.europe Pubmed central. 1989, 189 (3),225-234 .
  2. Takayuki Ezaki.;Yasuhiro Hashimoto.; Eiko Yabuuchi . Fluorometric Deoxyribonucleic Acid-Deoxyribonucleic Acid Hybridization in Microdilution Wells as an Alternative to Membrane Filter Hybridization in which Radioisotopes Are Used To Determine Genetic Relatedness among Bacterial Strains.international journal of systmatic and evolutionary microbiolgy. 1989, 39 (3),224-229 .