Melting point may be hazy or incomplete (forms polymer)
Enantiomeric excess
≥99.0% (U.S. Specification)
Proton NMR
Conforms to structure (U.S. Specification)
Optical Rotation
-10° to -11.5° (c=1 in DMF) (U.S. Specification)
Assay (HPLC)
≥97.0% (Non-U.S. Specification)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.