Thermo Scientific Chemicals

4-Amino-4'-chlorodiphenyl ether, 97%, Thermo Scientific Chemicals

Catalog number: B21083.18
50 g, Each
Thermo Scientific Chemicals

4-Amino-4'-chlorodiphenyl ether, 97%, Thermo Scientific Chemicals

Catalog number: B21083.18
50 g, Each
Quantity
Catalog number: B21083.18
also known as B21083-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-79-1
IUPAC Name
4-(4-chlorophenoxy)aniline
Molecular Formula
C12H10ClNO
InChI Key
YTISFYMPVILQRL-UHFFFAOYSA-N
SMILES
NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1
Appearance (Color)
Brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point
97.5-106.5?C

Description

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text