2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals
2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number B21087.22
also known as B21087-22
Price (USD)/ Each
141.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
141.00
Add to cart
2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals
Catalog numberB21087.22
Price (USD)/ Each
141.00
-
Add to cart
Chemical Identifiers
CAS589-10-6
IUPAC Name(2-bromoethoxy)benzene
Molecular FormulaC8H9BrO
InChI KeyJJFOBACUIRKUPN-UHFFFAOYSA-N
SMILESBrCCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)30.0-36.0?C
It is used in the synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Robin D. Clark, et al. Synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.J. Med. Chem.,198326(5), 657-661.
  2. Zhang-Lin Zhou, et al. 4-Hydroxy-1-[2-(4-hydroxyphenoxy)ethyl]-4-(4-methylbenzyl)piperidine:  A Novel, Potent, and Selective NR1/2B NMDA Receptor Antagonist.J. Med. Chem.,1999,42(15), 2993-3000.