Thermo Scientific™

Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals

Catalog number: B21101.03
1 g, Each
Thermo Scientific™

Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals

Catalog number: B21101.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21101.03
also known as B21101-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4559-70-0
IUPAC Name
(phenylphosphoroso)benzene
Molecular Formula
C12H11OP
InChI Key
ASUOLLHGALPRFK-UHFFFAOYSA-N
SMILES
O=P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or granules
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
48.0-58.0?C

Description

Used in the preparation of triarylphosphine oxides, and as asymmetric catalysts, preparation of alkenyldiphenylphosphine oxides, and Horner-Wittig reagents. It is a ligand for Buchwald-Hartwig Cross Coupling Reaction, Hydrophosphonylations and Suzuki-Miyaura Coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of triarylphosphine oxides, and as asymmetric catalysts, preparation of alkenyldiphenylphosphine oxides, and Horner-Wittig reagents. It is a ligand for Buchwald-Hartwig Cross Coupling Reaction, Hydrophosphonylations and Suzuki-Miyaura Coupling.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text