1-Trimethylsilyl-1,2,4-triazole, 95%, Thermo Scientific Chemicals
1-Trimethylsilyl-1,2,4-triazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Trimethylsilyl-1,2,4-triazole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21105.06
also known as B21105-06
Price (USD)/ Each
67.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
67.30
Add to cart
1-Trimethylsilyl-1,2,4-triazole, 95%, Thermo Scientific Chemicals
Catalog numberB21105.06
Price (USD)/ Each
67.30
-
Add to cart
Chemical Identifiers
CAS18293-54-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥94.0%
Proton NMRConforms to structure
Appearance (Color)Clear, colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only