N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals
N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B21107.14
also known as B21107-14
Price (USD)/ Each
168.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
168.00
Add to cart
N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals
Catalog numberB21107.14
Price (USD)/ Each
168.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Fmoc-L-alanine monohydrate
CAS207291-76-7
Melting Point∼130°C
Recommended StorageKeep cold
EINECS Number252-660-6
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only