Thermo Scientific Chemicals

Molecular sieves, 13X, 1.6-2.5mm (0.063-0.098in) beads, Thermo Scientific Chemicals

Catalog number: B21109.0I
5000 g, Each
Thermo Scientific Chemicals

Molecular sieves, 13X, 1.6-2.5mm (0.063-0.098in) beads, Thermo Scientific Chemicals

Catalog number: B21109.0I
5000 g, Each
Quantity
Catalog number: B21109.0I
also known as B21109-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63231-69-6
IUPAC Name
Molecular sieves 13X
MDL Number
MFCD00131613
Form
Beads
Capacity
CO₂ capacity: ≥11.5 lb per 100 lb (at 250 Torr, U.S. specification)
Crush Strength
Schleuniger crush strength (n=100) : ≥22N (non-U.S. specification)
Particle Size
≤5.0 wt% is >2.5mm (non-U.S. specification)
Crush Strength
≥5.0 lb (U.S. specification)

Description

They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text