2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
- Haselmeyer, A.; Zentek, J.; Chizzola, R. Effects of thyme as a feed additive in broiler chickens on thymol in gut contents, blood plasma, liver and muscle. J. Sci. Food Agric. 2015, 95 (3), 504-508.
- Kebede, B. T.; Grauwet, T.; Palmers, S.; Michiels, C.; Hendrickx, M.; Loey, A. V. Investigating chemical changes during shelf-life of thermal and high-pressure high-temperature sterilised carrot purees: A ‘fingerprinting kinetics’ approach. Food Chem. 2015, 185, 119-126.