Thermo Scientific Chemicals

Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals

Catalog number: B21112.09
10 g, Each
Thermo Scientific Chemicals

Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals

Catalog number: B21112.09
10 g, Each
Quantity
Catalog number: B21112.09
also known as B21112-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2071-20-7
IUPAC Name
[(diphenylphosphanyl)methyl]diphenylphosphane
Molecular Formula
C25H22P2
InChI Key
XGCDBGRZEKYHNV-UHFFFAOYSA-N
SMILES
C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Powder or crystalline powder of flakes
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
117-123?C

Description

It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text