Thermo Scientific Chemicals

Ethyl alpha-bromophenylacetate, 97%, Thermo Scientific Chemicals

Catalog number: B21114.30
250 g, Each
Thermo Scientific Chemicals

Ethyl alpha-bromophenylacetate, 97%, Thermo Scientific Chemicals

Catalog number: B21114.30
250 g, Each
Quantity
Catalog number: B21114.30
also known as B21114-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2882-19-1
IUPAC Name
ethyl 2-bromo-2-phenylacetate
Molecular Formula
C10H11BrO2
InChI Key
BKTKLDMYHTUESO-UHFFFAOYNA-N
SMILES
CCOC(=O)C(Br)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.5370-1.5420 @ 20?C
Form
Liquid

Description

α-Bromobenzeneacetic acid ethyl ester is used as an alkyl halide initiator in controlled/living radical polymerization of styrene and methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Bromobenzeneacetic acid ethyl ester is used as an alkyl halide initiator in controlled/living radical polymerization of styrene and methyl methacrylate.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text