Thermo Scientific Chemicals

Trimesitylphosphine, 98%, Thermo Scientific Chemicals

Catalog number: B21116.06
5 g, Each
Thermo Scientific Chemicals

Trimesitylphosphine, 98%, Thermo Scientific Chemicals

Catalog number: B21116.06
5 g, Each
Quantity
Catalog number: B21116.06
also known as B21116-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23897-15-6
IUPAC Name
tris(2,4,6-trimethylphenyl)phosphane
Molecular Formula
C27H33P
InChI Key
IDXDWPWXHTXJMZ-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(P(C2=C(C)C=C(C)C=C2C)C2=C(C)C=C(C)C=C2C)C(C)=C1
Appearance (Color)
White
Assay (GC)
>97.5%
Form
Powder
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure

Description

It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text