Thermo Scientific Chemicals

L-(+)-Ribose, 99%, Thermo Scientific Chemicals

Catalog number: B21117.03
1 g, Each
Thermo Scientific Chemicals

L-(+)-Ribose, 99%, Thermo Scientific Chemicals

Catalog number: B21117.03
1 g, Each
Quantity
Catalog number: B21117.03
also known as B21117-03
Price (USD)
Quantity
-
Have Questions?

Description

L-Ribose is a potential starting material for the synthesis of many L-nucleoside-based pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Ribose is a potential starting material for the synthesis of many L-nucleoside-based pharmaceutical compounds.

Solubility
Soluble in water (100 mg/ml).

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text