Thermo Scientific Chemicals

Bis(dimethylamino)dimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: B21119.22
100 g, Each
Thermo Scientific Chemicals

Bis(dimethylamino)dimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: B21119.22
100 g, Each
Quantity
Catalog number: B21119.22
also known as B21119-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3768-58-9
IUPAC Name
[(dimethylamino)dimethylsilyl]dimethylamine
Molecular Formula
C6H18N2Si
InChI Key
QULMGWCCKILBTO-UHFFFAOYSA-N
SMILES
CN(C)[Si](C)(C)N(C)C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Refractive Index
1.4155-1.4205 @ 20?C
Form
Liquid

Description

Bis(dimethylamino)-dimethyl silane has been used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterizing human electrogenic Na+-cotransporter isoform (hhNBC). It is also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes. It is also used in electronic film deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(dimethylamino)-dimethyl silane has been used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterizing human electrogenic Na+-cotransporter isoform (hhNBC). It is also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes. It is also used in electronic film deposition.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away. Protect from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text